Learn More
ALKBH5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16837-1-AP-150UL
 View more versions of this product
Product Code. 16886594
  • 4652.90 SEK / 150µL
Description
Dioxygenase that demethylates RNA by oxidative demethylation: specifically demethylates N(6)-methyladenosine (m6A) RNA, the most prevalent internal modification of messenger RNA (mRNA) in higher eukaryotes. Can also demethylate N(6)-methyladenosine in single-stranded DNA (in vitro). Requires molecular oxygen, alpha-ketoglutarate and iron. Demethylation of m6A mRNA affects mRNA processing and export. Required for spermatogenesis (By similarity).
Specifications
Specifications
ALKBH5
Polyclonal
Unconjugated
ALKBH5
ABH5, ALKBH5, OFOXD, OFOXD1
Rabbit
Antigen Affinity Chromatography
RUO
268420, 303193, 54890
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZKD3, Q3TSG4, Q6P6C2
Alkbh5
ALKBH5 Fusion Protein Ag10516
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ALKBH5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated