Learn More
Alkyd Resin, 70% Wt, Pure Solution, Fisher Chemical Product Code.: 10612251
Alkyd Resin, 70% Wt, Pure Solution, Fisher Chemical
Product Code. 10612251 Shop All Fisher Chemical Products
1L, Amber glass bottle
Click to view available options
1L, Amber glass bottle
Product Code. 10612251
Brand: Fisher Chemical A/1552/15
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Color Amber
Boiling Point 154°C
Packaging Amber glass bottle
MDL Number 1321655
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Alkyd resin
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Product Content Correction
Product Title
Alkyd Resin, 70% Wt, Pure Solution, Fisher Chemical >