Learn More
Alkyl-DHAP synthase/AGPS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21011-1-AP-150UL
Description
The Alkyl-DHAP synthase/AGPS antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Alkyl-DHAP synthase/AGPS. This antibody recognizes human antigen. The Alkyl-DHAP synthase/AGPS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Alkyl-DHAP synthase/AGPS | |
Polyclonal | |
Unconjugated | |
150 ÎĽL | |
8540 | |
IgG |
Western Blot, ELISA | |
560 ÎĽg/mL | |
Rabbit | |
Primary | |
Human |