missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1B-glycoprotein Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | alpha-1B-glycoprotein |
---|---|
Concentration | 0.15 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892704
|
Proteintech
14181-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882704
|
Proteintech
14181-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alpha-1-B glycoprotein (A1BG) which is a plasma protein with no known function belongs to the immunoglobulin superfamily. Although the human A1BG has been known for four decades, and the information about the amino acid sequence, chromosomal assignment, and even genetic polymorphism in different populations have been known, no biological function has been suggested. A1BG (Mr approximately equal to 63,000) consists of one polypeptide chain of 474 amino acids with four glucosamine oligosaccharides. The polypeptide has five intrachain disulfide bonds and consists of five repeating structural domains, each containing about 95 amino acids and one disulfide bond. Recently human cyctein-rich secretory protein 3 (CRISP-3) has been found to be a binding partner of A1B, suggesting the A1BG-CRISP-3 complex displays a protecting function from a potentially harmful effect of free CRISP-3 in circulation.Specifications
alpha-1B-glycoprotein | |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P04217, Q19LI2 | |
1, 117586 | |
alpha-1B-glycoprotein Fusion Protein Ag5414 | |
Primary | |
-20°C | |
A1BG |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A1B, A1BG, ABG, alpha 1 B glycoprotein, Alpha 1B glycoprotein, GAB, HYST2477 | |
A1BG | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
alpha-1B-glycoprotein Rabbit anti-Human, Mouse, Polyclonal, Proteintech