missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha galactosidase A Mouse anti-Human, Clone: 2B2C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66121-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predomitly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.Specifications
Alpha galactosidase A | |
Monoclonal | |
2.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06280 | |
GLA | |
Alpha galactosidase A Fusion Protein Ag7505 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
2B2C5 | |
Unconjugated | |
GLA | |
Alpha galactosidase A, galactosidase, alpha | |
Mouse | |
Protein A | |
RUO | |
2717 | |
-20°C | |
Liquid |