missing translation for 'onlineSavingsMsg'
Learn More
Skytec™ Alpha Sentinel Premium Reusable Half Face Mask Product Code.: 30242991
Only undefined left
Add to basket

Skytec™ Alpha Sentinel Premium Reusable Half Face Mask

Product Code. p-200104864
Small, Each
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30242991

Brand: Skytec™ ASRAS0001BD

Only undefined left
Add to basket
Only undefined left
Add to basket
This item is not returnable. View return policy

Alpha Sentinel Half Masks are ergonomically designed for optimum protection, comfort and PPE compatibility.

  • Well-balanced and lightweight (140 g), providing optimum wearer comfort
  • Available in three sizes, creating a tailored fit profile, superb fit and comfort for any workforce
  • Fully adjustable head harness, offering easy fitting and adjustment, with a robust and trusted fit when in use
  • Low profile shape to maximise the field of vision and allow exceptional compatibility with other PPE
  • Alpha Sentinel filters are fitted with a simple connection which easily clicks into position
  • Premium quality elastic strap provides a secure high comfort fit and retains mask positioning
  • The large exhalation valve reduces heat and moisture build-up, providing low breathing resistance
  • Easily cleaned and maintained with spare parts available
  • Quantitative face fit adapter available
  • Respirators are supplied with user seal check covers, which double as particulate retainers for combination filter

Specifications

Certifications/Compliance CE , UKCA , EN140
Color Black
Disposable Reusable
Filter Type HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA
Material Thermoplastic Elastomer
Product Type Half Mask Respirator
Size Small
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Filter dependent: Toxic Waste Disposal, Metal Cutting and Casting, Aerospace, Petrochemical
For Use With (Equipment) Alpha Sentinel Filters
Harness Material Thermoplastic Elastomer
Harness Style Standard
Latex Free Yes
Material Headstrap Polyester
Model Alpha Sentinel Half Mask
Neck Strap Polyester
Packaging Type Box
Quantity 16/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Skytec™ Alpha Sentinel Premium Reusable Half Face Mask >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.