missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Bound
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
8X-Large
Large
Medium
One Size
Small
X-Large
X-Small
Style:
W20B102
W20B103
W20B111
W20B113
W20B122
W20B162
Color:
White
White, Green
Description
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications

Specifications
Specifications
Size | 2X-Large |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Seam Type | Bound |
Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010,EN 13034:2005 + A1:2009,EN 1073-2:2002,EN 1149-5:2018,EN 14126:2003,EN ISO 13688:2013,DIN 32781:2010 |
For Use With (Application) | Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support and Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Crime Scene Investigation (CSI), Veterinary Services |
Weight (Metric) | 63 gsm |
Style | W20B162 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 2000 STANDARD Bound >
Spot an opportunity for improvement?Share a Content Correction