Learn More
Ansell AlphaTec™ 2300 STANDARD Bound
Chemical protective suits with a good protective barrier to numerous inorganic liquid chemicals including acids and bases.
Brand: Ansell YY23B-00111-03
5183.15 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Lightweight, yet relatively strong and durable
- Designed to protect, typical coverall features include respirator fit hood and a zip flap with self-adhesive tape closure
- Anti-static
Specifications
Medium | |
Yellow | |
EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 | |
65 gsm |
Polyethylene on Bicomponent Polypropylene, Polyethylene Non-Woven | |
Bound | |
Environmental Clean-up and Remediation, Assembly and Disassembly of Large Pipes, Transferring Liquids and Solids, Assembling and Mounting Components | |
Y23B111 |
Product Content Correction
Product Title
Ansell AlphaTec™ 2300 STANDARD Bound > Size: Medium, Style:Y23B111
Spot an opportunity for improvement?Share a Content Correction