missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16372-1-AP-20UL
This item is not returnable.
View return policy
Description
Transcriptional activator that acts at a palindromic recognition sequence to enhance the activity of the SV40 and TK promoters. Functions as a repressor with the prolactin promoter in vivo. May play a role in chondrocyte differentiation and may also influence cervix development.Specifications
ALX1 | |
Polyclonal | |
Unconjugated | |
ALX1 | |
ALX homeobox 1, ALX homeobox protein 1, ALX1, CART 1, CART1, Cartilage homeoprotein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216285, 25401, 8092 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15699, Q63087, Q8C8B0 | |
ALX1 | |
ALX1 Fusion Protein Ag9295 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ALX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction