missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMIGO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | AMIGO1 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891425
|
Proteintech
21874-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881425
|
Proteintech
21874-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Promotes growth and fasciculation of neurites from cultured hippocampal neurons. May be involved in fasciculation as well as myelination of developing neural axons. May have a role in regeneration as well as neural plasticity in the adult nervous system. May mediate homophilic as well as heterophilic cell-cell interaction and contribute to signal transduction through its intracellular domain.Specifications
AMIGO1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q80ZD7, Q80ZD8, Q86WK6 | |
229715, 295365, 57463 | |
AMIGO1 Fusion Protein Ag16226 | |
Primary | |
-20°C | |
AMIGO1 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALI2, Alivin 2, AMIGO, AMIGO 1, AMIGO1, Amphoterin induced protein 1, KIAA1163 | |
Amigo1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AMIGO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech