missing translation for 'onlineSavingsMsg'
Learn More

Amiselimod hydrochloride, MedChemExpress

MedChemExpress Amiselimod hydrochloride is a novel sphingosine 1-phosphate receptor-1 (S1P1) modulator, designed to reduce the bradycardia effects associated with fingolimod and other S1P receptor modulators.

Encompass_Preferred

575.75 SEK - 10441.17 SEK

Specifications

Chemical Name or Material Amiselimod hydrochloride
CAS 942398-84-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H31ClF3NO3
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16487156
Encompass_Preferred
View Documents
Medchem Express
HY-16734A
100 mg
10441.17 SEK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16459697
Encompass_Preferred
View Documents
Medchem Express
HY-16734A
5 mg
1447.13 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16457168
Encompass_Preferred
View Documents
Medchem Express
HY-16734A
10 mg
2411.90 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16441639
Encompass_Preferred
View Documents
Medchem Express
HY-16734A
1 mg
575.75 SEK
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16403197
Encompass_Preferred
View Documents
Medchem Express
HY-16734A
50 mg
7235.68 SEK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Amiselimod hydrochloride is a novel sphingosine 1-phosphate receptor-1 (S1P1) modulator, designed to reduce the bradycardia effects associated with fingolimod and other S1P receptor modulators.
Specifications

Specifications

Amiselimod hydrochloride
Off-White
C19H31ClF3NO3
DMSO : 110 mg/mL (265.76 mM; Need ultrasonic)
413.9
99.01%
Solid
942398-84-7
Research
MT-1303 hydrochloride
OCC(CCC1=CC=C(OCCCCCCC)C(C(F)(F)F)=C1)(N)CO.[H]Cl
413.9
Research
Cancer-programmed cell death
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Amiselimod hydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.