missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | AMOT |
---|---|
Concentration | 0.57 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883025
|
Proteintech
24550-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873025
|
Proteintech
24550-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMOT has two isoforms with MW 130 kDa (p130) and 80 kDa (p80). The p130 isoform can interact with F-actin. This antibody recognizes both p130 and p80.
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predomitly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
AMOT | |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q4VCS5 | |
154796 | |
AMOT Fusion Protein Ag19784 | |
Primary | |
-20°C | |
AMOT |
0.57 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AMOT, angiomotin, KIAA1071 | |
AMOT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AMOT Rabbit anti-Human, Polyclonal, Proteintech