missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphiregulin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Amphiregulin |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895494
|
Proteintech
16036-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885494
|
Proteintech
16036-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the epidermal growth factor family. It is an autocrine growth factor as well as a mitogen for astrocytes, Schwann cells, and fibroblasts. It is related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). This protein interacts with the EGF/TGF-alpha receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. This encoded protein is associated with a psoriasis-like skin phenotype.Specifications
Amphiregulin | |
Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
P15514 | |
374 | |
Amphiregulin Fusion Protein Ag8907 | |
Primary | |
-20°C | |
AREG |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
amphiregulin, AR, AREG, AREGB, CRDGF, SDGF | |
AREG | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Amphiregulin Rabbit anti-Human, Polyclonal, Proteintech