Learn More
AMPK Gamma 1 Mouse anti-Human, Unconjugated, Clone: 1E11D4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67182-1-IG-20UL
 View more versions of this product
Product Code. 16889675
  • 1645.54 SEK / 20µL
View Available Promotions
This item is not returnable. View return policy
Description
Description
The AMPK Gamma 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AMPK Gamma 1. This antibody recognizes human, mouse, rat antigen. The AMPK Gamma 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
AMPK Gamma 1
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1E11D4
Unconjugated
20 ÎĽL
5571
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AMPK Gamma 1 Mouse anti-Human, Unconjugated, Clone: 1E11D4, Proteintech > 20 ÎĽL