Learn More
AMPK Gamma 1 Mouse anti-Human, Unconjugated, Clone: 1E11D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67182-1-IG-150UL
Description
The AMPK Gamma 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AMPK Gamma 1. This antibody recognizes human, mouse, rat antigen. The AMPK Gamma 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
AMPK Gamma 1 | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1E11D4 | |
Unconjugated | |
150 ÎĽL | |
5571 | |
IgG1 |