missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 2/ANGPTL2 Rat anti-Mouse, Clone: 200001, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1444-SP

This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 2/ANGPTL2 Monoclonal specifically detects Angiopoietin-like Protein 2/ANGPTL2 in Mouse samples. It is validated for Western Blot.Specifications
Angiopoietin-like Protein 2/ANGPTL2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9R045 | |
ANGPTL2 | |
E. coli-derived recombinant mouse Angiopoietin-like 2 Pro23-His493 Accession # Q9R045 | |
25 ÎĽg | |
Primary | |
Detects mouse Angiopoietin-like 2 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant human (rh) Angiopoietin-like 2 but no cross-reactivity with rhAngiopoietin-1, -2, -4, rmAngiopoietin-3, rhAngiopoietin-like 1, or rhAngiopoietin-like 7. | |
Mouse | |
Purified |
Western Blot | |
200001 | |
Unconjugated | |
Lyophilized from a 0.2 ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 ÎĽm filtered solution in PBS. with No Preservative | |
angiopoietin-like 2, Angiopoietin-like protein 2, angiopoietin-related protein 2, ANGPTL2, ANGRP2, ARP2HARP, MGC8889 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23452 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Angiopoietin-like Protein 2/ANGPTL2 Rat anti-Mouse, Clone: 200001, R&D Systems™
Spot an opportunity for improvement?Share a Content Correction