Learn More
ANGPTL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12316-1-AP-20UL
 View more versions of this product
Product Code. 16839963
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
Specifications
Specifications
ANGPTL2
Polyclonal
Unconjugated
ANGPTL2
angiopoietin like 2, Angiopoietin like protein 2, Angiopoietin related protein 2, ANGPTL2, ARP2, HARP
Rabbit
Antigen Affinity Chromatography
RUO
23452, 26360
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9R045, Q9UKU9
ANGPTL2
ANGPTL2 Fusion Protein Ag2969
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANGPTL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated