missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23998-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ANKRD13A, also named as ANKRD13, a 590 amino acid protein, which contains two ANK repeats and four UIM (ubiquitin-interacting motif) repeats. ANKRD13A localizes in the cell membrane. ANKRD13A as ubiquitin-binding protein that specifically recognizes and binds ′Lys-63′-linked ubiquitin, not bind ′Lys-48′-linked ubiquitin. ANKRD13A positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.Specifications
ANKRD13A | |
Polyclonal | |
Unconjugated | |
ANKRD13A | |
ANKRD13, ANKRD13A, ankyrin repeat domain 13A, NY REN 25, Protein KE03 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
88455 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IZ07 | |
ANKRD13A | |
ANKRD13A Fusion Protein Ag21200 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |