missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ANKRD13A |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882595
|
Proteintech
23998-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16872595
|
Proteintech
23998-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ANKRD13A, also named as ANKRD13, a 590 amino acid protein, which contains two ANK repeats and four UIM (ubiquitin-interacting motif) repeats. ANKRD13A localizes in the cell membrane. ANKRD13A as ubiquitin-binding protein that specifically recognizes and binds ′Lys-63′-linked ubiquitin, not bind ′Lys-48′-linked ubiquitin. ANKRD13A positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.Specifications
ANKRD13A | |
Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8IZ07 | |
88455 | |
ANKRD13A Fusion Protein Ag21200 | |
Primary | |
-20°C | |
ANKRD13A |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ANKRD13, ANKRD13A, ankyrin repeat domain 13A, NY REN 25, Protein KE03 | |
ANKRD13A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |