missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25737-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody specifically recognizes the 70 kDa protein.
ANKRD13B (ankyrin repeat domain 13B) is an ubiquitin-binding protein belongs to the Ankrd 13 family. It specifically recognizes and binds ′Lys-63′-linked ubiquitin, and positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.Specifications
ANKRD13B | |
Polyclonal | |
Unconjugated | |
ANKRD13B | |
ANKRD13B, ankyrin repeat domain 13B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124930, 268445 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5F259, Q86YJ7 | |
ANKRD13B | |
ANKRD13B Fusion Protein Ag22732 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |