missing translation for 'onlineSavingsMsg'
Learn More

ANKRD13B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen ANKRD13B
Concentration 0.24 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16884295
View Documents
Proteintech
25737-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16874295
View Documents
Proteintech
25737-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody specifically recognizes the 70 kDa protein.

ANKRD13B (ankyrin repeat domain 13B) is an ubiquitin-binding protein belongs to the Ankrd 13 family. It specifically recognizes and binds ′Lys-63′-linked ubiquitin, and positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.
Specifications

Specifications

ANKRD13B
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Human
Q5F259, Q86YJ7
124930, 268445
ANKRD13B Fusion Protein Ag22732
Primary
-20°C
ANKRD13B
0.24 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ANKRD13B, ankyrin repeat domain 13B
ANKRD13B
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ANKRD13B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.