missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD50 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24032-1-AP-20UL
This item is not returnable.
View return policy
Description
Involved in the endosome-to-plasma membrane trafficking and recycling of SNX27-retromer-dependent cargo proteins, such as GLUT1.Specifications
ANKRD50 | |
Polyclonal | |
Unconjugated | |
ANKRD50 | |
ANKRD50, ankyrin repeat domain 50, KIAA1223 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57182 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ULJ7 | |
ANKRD50 | |
ANKRD50 Fusion Protein Ag21237 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANKRD50 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction