Learn More
ANKRD54 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25738-1-AP-150UL
 View more versions of this product
Product Code. 16894295
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Ankyrin (ANK) repeats mediate protein-protein interactions in diverse families of proteins. The number of ANK repeats in a protein can range from 2 to over 20. ANK repeats may occur in combinations with other types of domains. The structural repeat unit contains two anti-parallel helices and a beta-hairpin, with repeats stacked in a superhelical arrangement. LIAR, also known as ANKRD54, is a recently identified ANK repeat-containing protein that is predomitly expressed in tissues rich in ciliated cells, such as olfactory sensory neurons and is predicted to be important to cilia. At least three isoforms of LIAR are known to exist.
Specifications
Specifications
ANKRD54
Polyclonal
Unconjugated
ANKRD54
ANKRD54, ankyrin repeat domain 54
Rabbit
Antigen Affinity Chromatography
RUO
129138, 223690
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6NXT1, Q91WK7
ANKRD54
ANKRD54 Fusion Protein Ag22759
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANKRD54 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated