missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANO6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1395.65 SEK - 3774.95 SEK
Specifications
Antigen | ANO6 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
May act as a calcium-activated chloride channel.Specifications
ANO6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q4KMQ2, Q6P9J9 | |
105722, 196527 | |
ANO6 Fusion Protein Ag13937 | |
Primary | |
-20°C | |
ANO6 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ANO6, anoctamin 6, TMEM16F, Transmembrane protein 16F | |
Ano6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANO6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech