Learn More
anti-CCR2, APC, Clone: 475301, R&D Systems™
Rat Monoclonal Antibody has been used in 29 publications
Brand: R&D Systems FAB5538A-100

Description
CCR2 Monoclonal specifically detects CCR2 in Mouse samples. It is validated for Flow Cytometry.Specifications
CCR2 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CCR2 | |
L1.2 mouse pro-B cell line transfected with mouse CCR2 | |
100 Tests | |
729230 | |
Mouse | |
Purified |
Flow Cytometry | |
475301 | |
Flow Cytometry 10 uL/10^6 cells | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse CCR2 transfectants but not the parental cell line in flow cytometry. Does not detect HEK293 human embryonic kidney cell line transfected with mouse CCR5 in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.