Learn More
Histone H3 (pS28) Rat, Alexa Fluor 647, Clone: HTA28, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 558609
4811.79 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Histones are highly basic proteins that complex with DNA to form chromatin. Histone H3 is a ∽15kDa protein that is phosphorylated at serine 28 (S28), S10, and/or threonine 11 during mammalian cell mitosis and meiosis. The phosphorylation sites are located in the N-terminal tail, a region that is outside of the chromatin fiber and is thus accessible for interactions with agents that may regulate chromatin or specific gene activities. The phosphorylation states of the two serine sites during the cell cycle are highly regulated by Aurora B kinase and a PP1 phosphatase: S10 is in the phosphorylated state from late G2 phase to anaphase, while S28 is phosphorylated from prophase to anaphase. Furthermore, phosphorylation of histone H3 S28 may be mediated by other kinases in response to external stimuli. Evidence suggests that histone phosphorylation is involved in the regulation of chromosome condensation, cell division, and gene transcription. The HTA28 monoclonal antibody reacts with histone H3 phosphorylated at S28 in its N-terminal tail. It does not recognize the unphosphorylated protein.
Bioimaging

Specifications
Histone H3 (pS28) | |
Monoclonal | |
Alexa Fluor 647 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
In vitro Assay | |
HTA28 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Phosphorylated Human Histone H3 Peptide | |
100 Tests | |
Cell Biology | |
Bovine, Fly, Human, Mouse, Rat | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.