Learn More
IFN-Îł Mouse anti-Human, FITC, Clone: 4S.B3, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554551
4480.09 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Clone 4S.B3 reacts with the human form of interferon-Îł (IFN-Îł) detectable in the cytoplasm of a major subset of activated peripheral blood T lymphocytes. Clone 4S.B3 also cross-reacts with a cytoplasmic component of peripheral blood CD3+ lymphocytes of baboon, and both rhesus and cynomolgus macaque monkeys following five-hour treatment with phorbol myristic acetate (PMA) and Ca++ ionophore (A23187) in the presence of monensin. The staining pattern of 4S.B3 in CD3+ cells is similar to that observed with peripheral blood T lymphocytes from normal human donors. This reagent is useful for intracellular immunofluorescent staining for flow cytometric analysis to identify and enumerate IFN-Îł+ cells within a mixed cell population. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
IFN-Îł | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Partially purified human IFN-Îł from supernatants of human PBMC stimulated with Staphylococcus aureus | |
0.1 mg | |
Immunology | |
Human | |
IgG1 Îş |
Flow Cytometry | |
4S.B3 | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.