Learn More
anti-IgG2, κ, Clone: B81-3, BD
Brand:  BD Biosciences 559339
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15829718
  • 8410.00 SEK / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
The B81-3 antibody is specific for keyhole limpet hemocyanin. This antibody is routinely tested by LAL assay for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Blocking Assay, Co-Stimulation, Cytotoxicity Assay, Isotype Control
Specifications
Specifications
Blocking Assay, Functional Assay, Cytotoxicity Assay
1mg/mL
Hamster
0.5 mg
IgG2 κ
B81-3
Unconjugated
Affinity Purified
Undiluted at 4°C