Learn More
Lck (pY505) Mouse, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
3590.00 SEK - 7290.00 SEK
Specifications
Antigen | Lck (pY505) |
---|---|
Clone | 4 |
Concentration | 250ÎĽg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15836189
|
BD Biosciences
612390 |
50 ÎĽg |
3590.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15846189
|
BD Biosciences
612391 |
150 ÎĽg |
7290.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein tyrosine phosphorylation is an essential step in the signal transduction cascade leading to T cell antigen receptor (TCR) activation. Lck is a protein kinase and a member of the src family of cytoplasmic protein-tyrosine kinases (PTKs). Members of this family have several common features: 1) unique N-terminal domains, 2) attachment to cellular membranes through a myristylated N-terminus, and 3) homologous SH2, SH3, and catalytic domains. The unique N-terminal domain of Lck interacts with the cytoplasmic tails of the CD4 and CD8 cell surface
glycoproteins. CD4 and CD8 bind to surface MHC class II and class I molecules, respectively. Lck is regulated by both kinases and phosphatases. Autophosphorylation at Y394 leads to conformational changes in the catalytic domain, which induces kinase activity. Repression of Lck occurs via phosphorylation at Y505, located near the carboxy-terminus. Phosphorylation of this tyrosine site is mediated by the Csk family of PTKs. Upon phosphorylation at this site, Lck associates with the SH2 domain in the amino-terminus, thus keeping the protein biologically inactive. Lck activity and regulation is critical for activation and development of T cells.
Flow Cytometry, Intracellular Staining, Western Blotting
Specifications
Lck (pY505) | |
250ÎĽg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
4 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Human Lck (pY505) | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.