Learn More
Ly-51 Mouse anti-Mouse, PE, Clone: BP-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 553735
3807.06 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The BP-1 antibody reacts with an epitope of the 6C3/BP-1 (Ly-51) glycoprotein cell-surface differentiation antigen, which was originally identified on pre-B cell lymphomas (spontaneous and chemical- or retrovirus- transformed, in vitro and in vivo). 6C3/BP-1 is a homodimer cell-surface glycoprotein with 140kDa subunits which has been identified to possess aminopeptidase A (APA) activity. The same antigen is expressed at high levels on bone marrow stromal cell lines which support in vitro B lymphopoieses, on thymic dendritic cells and cortical epithelial cells, and on a wide variety of mouse and rat tissues known to possess APA activity. Subsets of normal bone marrow pre-B and B lymphocytes express low levels of Ly-51, which is rapidly up-regulated on the pre-B cells in the presence of IL-7. A role for the 6C3/BP-1 molecule in the IL-7-driven proliferation of B cell precursors has been postulated. However, B-cell abnormalities were not detected in Ly-51-deficient mice. Mature B lymphocytes, thymocytes, peripheral T lymphocytes, erythroid cells, and myeloid cells (with the exception of thymic dendritic cells) do not express Ly-51.
Host Species: Mouse
Clone: BP-1
Isotype: IgG2a Îş
Species Reactivity: Mouse
Immunogen: Abelson murine leukemia virus-transformed BALB/c mouse pre-B cell line 18.81
Flow Cytometry

Specifications
Ly-51 | |
Monoclonal | |
0.2mg/mL | |
6C3/BP-1 Antigen | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
BP-1 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Abelson murine leukemia virus-transformed BALB/c mouse pre-B cell line 18.81 | |
0.2 mg | |
Immunology | |
Mouse | |
IgG2a Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.