Learn More
anti-Caveolin-1, Biotin, Clone: 7C8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-615B
This item is not returnable.
View return policy
Description
Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
anti-Caveolin-1, Biotin, Clone: 7C8, Novus Biologicals™