Learn More
anti-NKG2A/CD159a, PE, Clone: 131411, R&D Systems™

Mouse Monoclonal Antibody has been used in 26 publications
Brand: R&D Systems FAB1059P-100

Description
NKG2A/CD159a/KLRC1 Monoclonal specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry.Specifications
NKG2A/CD159a/KLRC1 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
KLRC1 | |
BaF3 mouse pro-B cell line transfected with human NKG2A and CD94 | |
100 Tests | |
Primary | |
Recognizes the human NKG2A/CD94 heterodimer. It does not recognize the NKG2C/CD94 heterodimer or the CD94 homodimer. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
131411 | |
Flow Cytometry 10 uL/10^6 cells | |
CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3821 | |
Human | |
Purified |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.