Learn More
Invitrogen™ TCR V delta 2 Monoclonal Antibody (15D)
Mouse Monoclonal Antibody
Brand: Invitrogen™ TCR1732
Description
TCR1732 targets TCR V delta 2 in FACS, IHC (F), IP, WB and TCA applications and shows reactivity with Human samples. The TCR1732 immunogen is human TCR V delta 2.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
Specifications
TCR V delta 2 | |
Monoclonal | |
0.15 mg/mL | |
PBS with 0.5% BSA and 0.1% sodium azide | |
0 | |
Mouse | |
Protein A | |
RUO | |
6964 | |
-20°C | |
Liquid | |
Responsibly packaged |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation, Western Blot | |
15D | |
Unconjugated | |
TRD | |
TRD | |
Human TCR V delta 2. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Safety and Handling
- TCR V delta 2 Monoclonal Antibody
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AC406391 | Certificate of Analysis | 01/04/2025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.