missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3M1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12114-1-AP-20UL
This item is not returnable.
View return policy
Description
The AP3 complex is a heterotetramer composed of two large adaptins (AP3D1, AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2). The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. AP3M1 is highly homologous to AP3M2 but is the preferred binding partner to the YQRL motif from the cytosolic domain of TGN38. AP3M1 has also been shown to interact with the HIV-1 virulence protein Nef.Specifications
AP3M1 | |
Polyclonal | |
Unconjugated | |
AP3M1 | |
AP 3 complex subunit mu 1, AP3M1, Mu adaptin 3A, Mu3A adaptin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171126, 26985, 55946 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53676, Q9JKC8, Q9Y2T2 | |
Ap3m1 | |
AP3M1 Fusion Protein Ag2759 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AP3M1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction