missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOE Mouse anti-Human, Mouse, Clone: 1B2C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66830-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13.2.Specifications
APOE | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02649, P08226 | |
APOE | |
APOE Fusion Protein Ag28186 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1B2C9 | |
Unconjugated | |
APOE | |
AD2, Apo E, APOE, apolipoprotein E, LPG | |
Mouse | |
Protein G | |
RUO | |
11816, 348 | |
-20°C | |
Liquid |