Learn More
APOF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16608-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene is one of the minor apolipoproteins found in plasma. This protein forms complexes with lipoproteins and may be involved in transport and/or esterification of cholesterol.Specifications
APOF | |
Polyclonal | |
Unconjugated | |
APOF | |
Apo F, APOF, apolipoprotein F, LTIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
319 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13790 | |
APOF | |
APOF Fusion Protein Ag9824 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
APOF Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction