missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein H Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4D9A4, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen Apolipoprotein H
Clone 4D9A4
Concentration 0.78 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897615
View Documents
Proteintech
66074-1-IG-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16887615
View Documents
Proteintech
66074-1-IG-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
Specifications

Specifications

Apolipoprotein H
0.78 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Anticardiolipin cofactor, APC inhibitor, Apo H, APOH, Apolipoprotein H, B2G1, B2GPI, Beta 2 glycoprotein 1, Beta 2 glycoprotein I, Beta(2)GPI, BG
Apoh
IgG1
Protein G
Antibody
4D9A4
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Rat, Pig, Human, Mouse
P02749, P26644, Q01339
100522100, 11818, 287774, 350
Apolipoprotein H Fusion Protein Ag17915
Primary
-20°C
APOH
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Apolipoprotein H Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4D9A4, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.