missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AQP1 Mouse anti-Human, Porcine, Clone: 2E5D6, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | AQP1 |
---|---|
Clone | 2E5D6 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889015
|
Proteintech
66805-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16879015
|
Proteintech
66805-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.Specifications
AQP1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AQP 1, AQP CHIP, AQP1, Aquaporin 1, Aquaporin CHIP, CHIP28, CO, Urine water channel | |
AQP1 | |
IgG1 | |
Protein G | |
Antibody |
2E5D6 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Porcine | |
P29972, Q6PQZ1 | |
358, 407773 | |
AQP1 Fusion Protein Ag14093 | |
Primary | |
-20°C | |
AQP1 |