missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AQP1 Mouse anti-Human, Porcine, Clone: 2E5D6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66805-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.Specifications
AQP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29972, Q6PQZ1 | |
AQP1 | |
AQP1 Fusion Protein Ag14093 | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2E5D6 | |
Unconjugated | |
AQP1 | |
AQP 1, AQP CHIP, AQP1, Aquaporin 1, Aquaporin CHIP, CHIP28, CO, Urine water channel | |
Mouse | |
Protein G | |
RUO | |
358, 407773 | |
-20°C | |
Liquid |