missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARD1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14803-1-AP-20UL
This item is not returnable.
View return policy
Description
N-alpha-acetylation is one of the most common protein modifications that occurs during protein synthesis and involves the transfer of an acetyl group from acetyl-coenzyme A to the protein alpha-amino group. ARD1A, together with NATH.Specifications
ARD1A | |
Polyclonal | |
Unconjugated | |
NAA10 | |
ARD1, ARD1A, DXS707, NAA10, TE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363518, 56292, 8260 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P41227, Q9QY36 | |
Naa10 | |
ARD1A Fusion Protein Ag6519 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARD1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction