missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11243-1-AP-20UL
This item is not returnable.
View return policy
Description
Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
ARHGEF18 | |
Polyclonal | |
Unconjugated | |
ARHGEF18 | |
ARHGEF18, KIAA0521, p114 Rho GEF, P114 RhoGEF, p114RhoGEF, SA RhoGEF, Septin associated RhoGEF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102098, 23370 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P9R4, Q6ZSZ5 | |
Arhgef18 | |
ARHGEF18 Fusion Protein Ag1749 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARHGEF18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction