missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20506-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that contains an armadillo repeat and transmembrane domain. The encoded protein decreases the transcriptional activity of the tumor suppressor protein p53 through direct interaction with the DNA-binding domain of p53, and may play a role in cell growth and survival. Upregulation of this gene may play a role in hepatocellular carcinoma. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 3.Specifications
ARMC10 | |
Polyclonal | |
Unconjugated | |
ARMC10 | |
armadillo repeat containing 10, ARMC10, PNAS 112, SVH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296758, 67211, 83787 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B1WBW4, Q8N2F6, Q9D0L7 | |
Armc10 | |
ARMC10 Fusion Protein Ag14313 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARMC10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction