missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24029-1-AP-20UL
This item is not returnable.
View return policy
Description
ARMC7 is a protein coding gene.Specifications
ARMC7 | |
Polyclonal | |
Unconjugated | |
ARMC7 | |
armadillo repeat containing 7, ARMC7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79637 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H6L4 | |
ARMC7 | |
ARMC7 Fusion Protein Ag21222 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARMC7 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction