Learn More
ARMC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12653-1-AP-150UL
 View more versions of this product
Product Code. 16880484
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
ARMC8 is a protein coding gene.
Specifications
Specifications
ARMC8
Polyclonal
Unconjugated
ARMC8
armadillo repeat containing 8, ARMC8, HSPC056, S863 2
Rabbit
Antigen Affinity Chromatography
RUO
25852, 315949, 74125
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IUR7, Q9DBR3
Armc8
ARMC8 Fusion Protein Ag3374
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARMC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated