Learn More
AS3MT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27270-1-AP-150UL
 View more versions of this product
Product Code. 16875525
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Specifications
AS3MT
Polyclonal
Unconjugated
AS3MT
Arsenite methyltransferase, AS3MT, CYT19
Rabbit
Antigen Affinity Chromatography
RUO
57344, 57412
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q91WU5, Q9HBK9
As3mt
AS3MT Fusion Protein Ag26194
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AS3MT Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated