Learn More
AS3MT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27270-1-AP-150UL
This item is not returnable.
View return policy
Specifications
AS3MT | |
Polyclonal | |
Unconjugated | |
AS3MT | |
Arsenite methyltransferase, AS3MT, CYT19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57344, 57412 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91WU5, Q9HBK9 | |
As3mt | |
AS3MT Fusion Protein Ag26194 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
AS3MT Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction