Learn More
ASB14 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25300-1-AP-20UL
 View more versions of this product
Product Code. 16883885
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
ASB14 gene ontology annotations related to this gene include intracellular signal transduction; protein ubiquitination.
Specifications
Specifications
ASB14
Polyclonal
Unconjugated
ASB14
ASB 14, ASB14
Rabbit
Antigen Affinity Chromatography
RUO
142686, 142687
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6NK59, Q8C6Y6
ASB14
ASB14 Fusion Protein Ag18147
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ASB14 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated