Learn More
ASCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12301-1-AP-150UL
 View more versions of this product
Product Code. 16889933
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a subunit of the activating signal cointegrator 1 (ASC-1) complex. The ASC-1 complex is a transcriptional coactivator that plays an important role in gene transactivation by multiple transcription factors including activating protein 1 (AP-1), nuclear factor kappa-B (NF-kB) and serum response factor (SRF). The encoded protein contains an N-terminal KH-type RNA-binding motif which is required for AP-1 transactivation by the ASC-1 complex. Mutations in this gene are associated with Barrett esophagus and esophageal adenocarcinoma. Alternatively spliced transcripts encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
ASCC1
Polyclonal
Unconjugated
ASCC1
ASC 1 complex subunit p50, ASC1p50, ASCC1, CGI 18, RP11 150D20.4, Trip4 complex subunit p50
Rabbit
Antigen Affinity Chromatography
RUO
294512, 51008, 69090
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N9N2, Q9D8Z1
ASCC1
ASCC1 Fusion Protein Ag2946
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ASCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated