Learn More
ASCT2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ASCT2 |
---|---|
Concentration | 433 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899874
|
Proteintech
20350-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889874
|
Proteintech
20350-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ASCT2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ASCT2. This antibody recognizes human, mouse, antigen. The ASCT2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ASCT2 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse | |
IgG |
433 μg/mL | |
Polyclonal | |
Rabbit | |
6510 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.