Learn More
ASCT2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20350-1-AP-150UL
Description
The ASCT2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ASCT2. This antibody recognizes human, mouse, antigen. The ASCT2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ASCT2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
6510 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
433 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |